2-Bromo-5-fluoro-4-(trifluoromethyl)phenylacetic acid
Supplier: Apollo Scientific
PC303555-1G PC303555-5G
APOSPC303555-1GEA 430 EUR
APOSPC303555-1G APOSPC303555-5G
2-Bromo-5-fluoro-4-(trifluoromethyl)phenylacetic acid
2-Bromo-5-fluoro-4-(trifluoromethyl)phenylacetic acid
Formula: C₉H₅BrF₄O₂
MW: 301.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640133
CAS Number: 2385550-69-4