2-Bromo-5-fluoro-4-(trifluoromethyl)benzoyl chloride
Supplier: Apollo Scientific
PC303558-1G PC303558-5G
APOSPC303558-1GEA 244 EUR
APOSPC303558-1G APOSPC303558-5G
2-Bromo-5-fluoro-4-(trifluoromethyl)benzoyl chloride
2-Bromo-5-fluoro-4-(trifluoromethyl)benzoyl chloride
Formula: C₈H₂BrClF₄O
MW: 305.451 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640136
CAS Number: 2168880-84-8