2-Bromo-5-fluoro-4-(trifluoromethyl)benzonitrile
Supplier: Apollo Scientific
PC303560-1G PC303560-5G
APOSPC303560-1GEA 0 EUR
APOSPC303560-1G APOSPC303560-5G
2-Bromo-5-fluoro-4-(trifluoromethyl)benzonitrile
2-Bromo-5-fluoro-4-(trifluoromethyl)benzonitrile
Formula: C₈H₂BrF₄N
MW: 268.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640138
CAS Number: 2090465-86-2