7H-Perfluoroheptanoic acid 97%
Supplier: Apollo Scientific
Danger
PC3131-100G PC3131-25G
APOSPC3131-100GEA 102 EUR
APOSPC3131-100G APOSPC3131-25G
7H-Perfluoroheptanoic acid 97%
7H-Perfluoroheptanoic acid
Formula: C₇H₂F₁₂O₂
Boiling Pt: 175 °C (42 mm Hg)
Melting Pt: 34 °C
Density: 1.792 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00054694
CAS Number: 1546-95-8
EINECS: 216-283-0
UN: 3261
ADR: 8,II