Ethyl 2-{2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetate
Supplier: Apollo Scientific
PC32561-1G PC32561-5G
APOSPC32561-1GEA 132 EUR
APOSPC32561-1G APOSPC32561-5G
Ethyl 2-{2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetate
Ethyl 2-{2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetate
Formula: C₁₄H₁₂F₃NO₂S
Melting Pt: 37.8…40.6 °C
Storage Temperature: Ambient
MDL Number: MFCD01765079
CAS Number: 680215-92-3