Methyl 2-fluoro-4-methyl-5-nitrobenzoate
Supplier: Apollo Scientific
Warning
PC32909-1G PC32909-250MG PC32909-5G
APOSPC32909-1GEA 140 EUR
APOSPC32909-1G APOSPC32909-250MG APOSPC32909-5G
Methyl 2-fluoro-4-methyl-5-nitrobenzoate
Methyl 2-fluoro-4-methyl-5-nitrobenzoate
Formula: C₉H₈FNO₄
MW: 213.163 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23863743
CAS Number: 753924-48-0