2-[Bromo(difluoro)methyl]-4-(trifluoromethyl)-1H-benzimidazole
Supplier: Apollo Scientific
Warning
PC32923-1G PC32923-250MG PC32923-5G
APOSPC32923-1GEA 77 EUR
APOSPC32923-1G APOSPC32923-250MG APOSPC32923-5G
2-[Bromo(difluoro)methyl]-4-(trifluoromethyl)-1H-benzimidazole
2-[Bromo(difluoro)methyl]-4-(trifluoromethyl)-1H-benzimidazole
Formula: C₉H₄BrF₅N₂
MW: 315.037 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31922506
CAS Number: 1353778-48-9