2-[Bromo(difluoro)methyl]-5-(trifluoromethoxy)-1H-benzimidazole
Supplier: Apollo Scientific
Warning
PC32926-1G PC32926-250MG PC32926-5G
APOSPC32926-1GEA 78.3 EUR
APOSPC32926-1G APOSPC32926-250MG APOSPC32926-5G
2-[Bromo(difluoro)methyl]-5-(trifluoromethoxy)-1H-benzimidazole
2-[Bromo(difluoro)methyl]-5-(trifluoromethoxy)-1H-benzimidazole
Formula: C₉H₄BrF₅N₂O
MW: 331.037 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31922509
CAS Number: 113638-38-3