2-(Difluoromethyl)-4-chloro-1H-benzimidazole
Supplier: Apollo Scientific
Warning
PC32936-1G PC32936-250MG
APOSPC32936-1GEA 186 EUR
APOSPC32936-1G APOSPC32936-250MG
2-(Difluoromethyl)-4-chloro-1H-benzimidazole
2-(Difluoromethyl)-4-chloro-1H-benzimidazole
Formula: C₈H₅ClF₂N₂
MW: 202.589 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20390926
CAS Number: 1342424-21-8