Methyl 3,6-difluoro-2-(trifluoromethyl)benzoate 95%
Supplier: Apollo Scientific
Warning
PC32983-1G PC32983-250MG PC32983-5G
APOSPC32983-1GEA 522 EUR
APOSPC32983-1G APOSPC32983-250MG APOSPC32983-5G
Methyl 3,6-difluoro-2-(trifluoromethyl)benzoate 95%
Methyl 3,6-difluoro-2-(trifluoromethyl)benzoate
Formula: C₉H₅F₅O₂
MW: 240.127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32066744
CAS Number: 2366994-08-1