2,3-Dichloro-4-fluoronitrobenzene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2,3-Dichloro-1-fluoro-4-nitrobenzene
PC3682-1G PC3682-25G PC3682-5G
APOSPC3682-1GEA 21.9 EUR
APOSPC3682-1G APOSPC3682-25G APOSPC3682-5G
2,3-Dichloro-4-fluoronitrobenzene 97%
2,3-Dichloro-4-fluoronitrobenzene
Formula: C₆H₂Cl₂FNO₂
Density: 1.6 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD09763598
CAS Number: 36556-51-1