Methyl 6-fluoro-4-(isopropylamino)pyridine-3-carboxylate
Supplier: Apollo Scientific
Warning
PC400035-1G PC400035-250MG
APOSPC400035-1GEA 644 EUR
APOSPC400035-1G APOSPC400035-250MG
Methyl 6-fluoro-4-(isopropylamino)pyridine-3-carboxylate
Methyl 6-fluoro-4-(isopropylamino)pyridine-3-carboxylate
Formula: C₁₀H₁₃FN₂O₂
MW: 212.221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31536750
CAS Number: 2187434-95-1