1-(2,2-Dibromovinyl)-2-fluorobenzene
Supplier: Apollo Scientific
Danger
PC400040-250MG PC400040-1G
APOSPC400040-250MGEA 102 EUR
APOSPC400040-250MG APOSPC400040-1G
1-(2,2-Dibromovinyl)-2-fluorobenzene
1-(2,2-Dibromovinyl)-2-fluorobenzene
Formula: C₈H₅Br₂F
MW: 279.932 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30537997
CAS Number: 401514-42-9
UN: 2811
ADR: 6.1 III