2-(Difluoromethyl)-5-nitropyridin-4-ol
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Difluoromethyl)-4-hydroxy-5-nitropyridine
PC400192-1G PC400192-250MG PC400192-5G
APOSPC400192-1GEA 857 EUR
APOSPC400192-1G APOSPC400192-250MG APOSPC400192-5G
2-(Difluoromethyl)-5-nitropyridin-4-ol
2-(Difluoromethyl)-5-nitropyridin-4-ol
Formula: C₆H₄F₂N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD25481266
CAS Number: 1805314-41-3