cis-tert-Butyl 3-(4-trifluoromethylphenyl)-aziridine-2-carboxylate
Supplier: Apollo Scientific
PC404511-1G PC404511-250MG
APOSPC404511-1GEA 525 EUR
APOSPC404511-1G APOSPC404511-250MG
cis-tert-Butyl 3-(4-trifluoromethylphenyl)-aziridine-2-carboxylate
cis-tert-Butyl 3-(4-trifluoromethylphenyl)-aziridine-2-carboxylate
Formula: C₁₄H₁₆F₃NO₂
MW: 287.278 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26936313
CAS Number: 1980007-28-0