3-[4-(Trifluoromethyl)phenyl]propanal
Supplier: Apollo Scientific
Warning
Synonyms: 3-[4-(Trifluoromethyl)phenyl]propionaldehyde, 4-(3-Oxoprop-1-yl)benzotrifluoride
PC4059-100MG PC4059-250MG
APOSPC4059-100MGEA 63.8 EUR
APOSPC4059-100MG APOSPC4059-250MG
3-[4-(Trifluoromethyl)phenyl]propanal
3-[4-(Trifluoromethyl)phenyl]propanal
Formula: C₁₀H₉F₃O
MW: 202.173 g/mol
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD07772919
CAS Number: 166947-09-7