6,6-Difluorospiro[2.5]octane-1-carboxylic acid
Supplier: Apollo Scientific
PC411286-1G PC411286-250MG PC411286-500MG
APOSPC411286-1GEA 1830 EUR
APOSPC411286-1G APOSPC411286-250MG APOSPC411286-500MG
6,6-Difluorospiro[2.5]octane-1-carboxylic acid
6,6-Difluorospiro[2.5]octane-1-carboxylic acid
Formula: C₉H₁₂F₂O₂
MW: 190.187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27995747
CAS Number: 1447943-53-4