Ethyl 6,6-difluorospiro[2.5]octane-1-carboxylate
Supplier: Apollo Scientific
PC411287-1G PC411287-250MG PC411287-500MG
APOSPC411287-1GEA 811 EUR
APOSPC411287-1G APOSPC411287-250MG APOSPC411287-500MG
Ethyl 6,6-difluorospiro[2.5]octane-1-carboxylate
Ethyl 6,6-difluorospiro[2.5]octane-1-carboxylate
Formula: C₁₁H₁₆F₂O₂
MW: 218.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30471658
CAS Number: 1447942-88-2