4-(Chlorodifluoromethoxy)aniline 96%
Supplier: Apollo Scientific
|
Warning
|
PC4124-1G
PC4124-5G
PC4124-250MG
APOSPC4124-1GEA
24.4
EUR
APOSPC4124-1G
APOSPC4124-5G
APOSPC4124-250MG
4-(Chlorodifluoromethoxy)aniline 96%
4-(Chlorodifluoromethoxy)aniline
Formula:
C₇H₆ClF₂NO Storage Temperature: Ambient |
MDL Number:
MFCD01820809 CAS Number: 39065-95-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...