2,2-Difluoro-7-azaspiro[3.5]nonane hydrochloride 97%
Supplier: Apollo Scientific
Warning
PC420025-1G PC420025-500MG
APOSPC420025-1GEA 621 EUR
APOSPC420025-1G APOSPC420025-500MG
2,2-Difluoro-7-azaspiro[3.5]nonane hydrochloride 97%
2,2-Difluoro-7-azaspiro[3.5]nonane hydrochloride
Formula: C₈H₁₃F₂N·ClH
MW: 197.653 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22683336
CAS Number: 1523618-02-1