2-Bromo-5-(trifluoromethoxy)pyridine
Supplier: Apollo Scientific
Danger
PC421021-1G PC421021-500MG PC421021-5G
APOSPC421021-1GEA 190 EUR
APOSPC421021-1G APOSPC421021-500MG APOSPC421021-5G
2-Bromo-5-(trifluoromethoxy)pyridine
2-Bromo-5-(trifluoromethoxy)pyridine
Formula: C₆H₃BrF₃NO
MW: 241.993 g/mol
Boiling Pt: 75 °C (15 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD10698596
CAS Number: 888327-36-4
UN: 2810
ADR: 6.1 III