2-Chloro-4-trifluoromethoxy-benzaldehyde 95%
Supplier: Apollo Scientific
Synonyms: 2-Chloro-4-(trifluoromethoxy)benzaldehyde
PC421043-1G PC421043-250MG PC421043-5G
APOSPC421043-1GEA 224 EUR
APOSPC421043-1G APOSPC421043-250MG APOSPC421043-5G
2-Chloro-4-trifluoromethoxy-benzaldehyde 95%
2-Chloro-4-trifluoromethoxy-benzaldehyde
Formula: C₈H₄ClF₃O₂
MW: 224.564 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18393768
CAS Number: 1079351-20-4