4-(Trifluoromethyl)-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine 95%
Supplier: Apollo Scientific
Warning
PC42589-1G PC42589-250MG
APOSPC42589-1GEA 567 EUR
APOSPC42589-1G APOSPC42589-250MG
4-(Trifluoromethyl)-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine 95%
4-(Trifluoromethyl)-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine
Formula: C₈H₇F₃N₂
MW: 188.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31812114
CAS Number: 2366994-59-2