2,4,6-Tris(difluoromethyl)-1,3,5-triazine
Supplier: Apollo Scientific
Warning
Synonyms: 2,4,6-Tris(difluoromethyl)-s-triazine
PC4281-1G PC4281-5G
APOSPC4281-1GEA 152 EUR
APOSPC4281-1G APOSPC4281-5G
2,4,6-Tris(difluoromethyl)-1,3,5-triazine
2,4,6-Tris(difluoromethyl)-1,3,5-triazine
Formula: C₆H₃F₆N₃
Boiling Pt: 72…73 °C (20 mm Hg)
Melting Pt: 31…34 °C
Density: 1.5973 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD08059507
CAS Number: 369-22-2