3-(Trifluoromethyl)phenylurea 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-(Carbamoylamino)benzotrifluoride, 3-Ureidobenzotrifluoride
PC4344-10G PC4344-1G PC4344-50G PC4344-100G
APOSPC4344-10GEA 29 EUR
APOSPC4344-10G APOSPC4344-1G APOSPC4344-50G APOSPC4344-100G
3-(Trifluoromethyl)phenylurea 98%
3-(Trifluoromethyl)phenylurea
Formula: C₈H₇F₃N₂O
Melting Pt: 101…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00025424
CAS Number: 13114-87-9