2-[4-Bromo-2-(trifluoromethyl)phenyl]amino-4-(2-hydroxyphenyl)-1,3-thiazole hydrobromide 97%
Supplier: Apollo Scientific
Warning
PC4485-1G PC4485-5G
APOSPC4485-1GEA 279 EUR
APOSPC4485-1G APOSPC4485-5G
2-[4-Bromo-2-(trifluoromethyl)phenyl]amino-4-(2-hydroxyphenyl)-1,3-thiazole hydrobromide 97%
2-[4-Bromo-2-(trifluoromethyl)phenyl]amino-4-(2-hydroxyphenyl)-1,3-thiazole hydrobromide
Formula: C₁₆H₁₀BrF₃N₂OS·BrH
MW: 496.14 g/mol
Melting Pt: 235…240 °C
Storage Temperature: Ambient
MDL Number: MFCD08458095
CAS Number: 1210389-46-0