4,6-Bis(hydroxy)-4.6-bis(trifluoromethyl)tetrahydropyrimidine-2-thione
Supplier: Apollo Scientific
PC450037-100MG PC450037-250MG
APOSPC450037-100MGEA 116 EUR
APOSPC450037-100MG APOSPC450037-250MG
4,6-Bis(hydroxy)-4.6-bis(trifluoromethyl)tetrahydropyrimidine-2-thione
4,6-Bis(hydroxy)-4.6-bis(trifluoromethyl)tetrahydropyrimidine-2-thione
Formula: C₆H₆F₆N₂O₂S
MW: 284.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26408204
CAS Number: 1820712-27-3