Ethyl 2,2-difluoro-3-hydroxyhexanoate
Supplier: Apollo Scientific
PC450060-1G PC450060-250MG
APOSPC450060-1GEA 865 EUR
APOSPC450060-1G APOSPC450060-250MG
Ethyl 2,2-difluoro-3-hydroxyhexanoate
Ethyl 2,2-difluoro-3-hydroxyhexanoate
Formula: C₈H₁₄F₂O₃
MW: 196.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16749399
CAS Number: 156544-72-8