2,2-Difluorocyclooctan-1-one
Supplier: Apollo Scientific
PC450061-100MG PC450061-250MG
APOSPC450061-100MGEA 147 EUR
APOSPC450061-100MG APOSPC450061-250MG
2,2-Difluorocyclooctan-1-one
2,2-Difluorocyclooctan-1-one
Formula: C₈H₁₂F₂O
MW: 162.177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24698073
CAS Number: 1435806-60-2