Perfluoro(2,5,8-trimethyl-3,6,9-trioxadodecan)amide
Supplier: Apollo Scientific
PC450122-1G PC450122-5G
APOSPC450122-1GEA 241 EUR
APOSPC450122-1G APOSPC450122-5G
Perfluoro(2,5,8-trimethyl-3,6,9-trioxadodecan)amide
Perfluoro(2,5,8-trimethyl-3,6,9-trioxadodecan)amide
Formula: C₁₂H₂F₂₃NO₄
Boiling Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD19442118
CAS Number: 51445-02-4