2,5-Bis(trifluoromethyl)-1H-benzimidazolyl-6-carboxylic acid
Supplier: Apollo Scientific
Warning
PC450136-1G PC450136-250MG
APOSPC450136-1GEA 611 EUR
APOSPC450136-1G APOSPC450136-250MG
2,5-Bis(trifluoromethyl)-1H-benzimidazolyl-6-carboxylic acid
2,5-Bis(trifluoromethyl)-1H-benzimidazolyl-6-carboxylic acid
Formula: C₁₀H₄F₆N₂O₂
MW: 298.141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18072173
CAS Number: 1309602-77-4