5,6-Dichloro-2-(trifluoromethyl)-1H-benzimidazole
Supplier: Apollo Scientific
Danger
PC450181-1G PC450181-5G
APOSPC450181-1GEA 259 EUR
APOSPC450181-1G APOSPC450181-5G
5,6-Dichloro-2-(trifluoromethyl)-1H-benzimidazole
5,6-Dichloro-2-(trifluoromethyl)-1H-benzimidazole
Formula: C₈H₃Cl₂F₃N₂
MW: 255.024 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01658509
CAS Number: 2338-25-2
UN: 2811
ADR: 6.1 II