N-Cyclohexyl-2,2,5,5-tetrakis(trifluoromethyl)-1,3-dioxolan-4-imine 95%
Supplier: Apollo Scientific
PC450488-1G PC450488-250MG
APOSPC450488-1GEA 919 EUR
APOSPC450488-1G APOSPC450488-250MG
N-Cyclohexyl-2,2,5,5-tetrakis(trifluoromethyl)-1,3-dioxolan-4-imine 95%
N-Cyclohexyl-2,2,5,5-tetrakis(trifluoromethyl)-1,3-dioxolan-4-imine
Formula: C₁₃H₁₁F₁₂NO₂
MW: 441.213 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29765768
CAS Number: 5771-96-0