1H-Heptafluoropropane (FC-227ca) 97%
Supplier: Apollo Scientific
Warning
PC4505-100G PC4505-25G PC4505-5G
APOSPC4505-100GEA 1320 EUR
APOSPC4505-100G APOSPC4505-25G APOSPC4505-5G
1H-Heptafluoropropane (FC-227ca) 97%
1H-Heptafluoropropane (FC-227ca)
Formula: C₃HF₇
Boiling Pt: –16 °C
Melting Pt: –129.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00236614
CAS Number: 2252-84-8
UN: 3296
ADR: 2.2,