trans-4,4,4-Trifluoro-2-buten-1-ol
Supplier: Apollo Scientific
Warning
Synonyms: (E)-4,4,4-Trifluorobut-2-en-1-ol
PC450522-1G PC450522-250MG
APOSPC450522-1GEA 725 EUR
APOSPC450522-1G APOSPC450522-250MG
trans-4,4,4-Trifluoro-2-buten-1-ol
trans-4,4,4-Trifluoro-2-buten-1-ol
Formula: C₄H₅F₃O
Boiling Pt: 128…129 °C (760 mm Hg)
Density: 1.256 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00077603
CAS Number: 83706-94-9
UN: 1987
ADR: 3,III