2-Chlorobutylsulphurpentafluoride

Supplier: Apollo Scientific
Warning

PC450567-1G PC450567-250MG
APOSPC450567-1GEA 690 EUR
APOSPC450567-1G APOSPC450567-250MG
2-Chlorobutylsulphurpentafluoride
2-Chlorobutylsulphurpentafluoride
Formula: C₄H₈ClF₅S
Storage Temperature: Ambient
MDL Number: MFCD31695424
CAS Number: 2244079-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR