2-Amino-5-(trifluoromethyl)nicotinonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-5-(trifluoromethyl)pyridine-3-carbonitrile, 2-Amino-3-cyano-5-(trifluoromethyl)pyridine
PC48319-1G PC48319-500MG
APOSPC48319-1GEA 568 EUR
APOSPC48319-1G APOSPC48319-500MG
2-Amino-5-(trifluoromethyl)nicotinonitrile
2-Amino-5-(trifluoromethyl)nicotinonitrile
Formula: C₇H₄F₃N₃
MW: 187.122 g/mol
Melting Pt: 170…172 °C
Storage Temperature: Ambient
MDL Number: MFCD18261102
CAS Number: 1211514-97-4
UN: 3439
ADR: 6.1 III