4-Fluoro-2-nitrobenzyl bromide 98%
Supplier: Apollo Scientific
Danger
Synonyms: 1-(Bromomethyl)-4-fluoro-2-nitrobenzene, α-Bromo-4-fluoro-2-nitrotoluene
PC48320-10G PC48320-50G PC48320-100G PC48320-5G PC48320-25G
APOSPC48320-10GEA 64 EUR
APOSPC48320-10G APOSPC48320-50G APOSPC48320-100G APOSPC48320-5G APOSPC48320-25G
4-Fluoro-2-nitrobenzyl bromide 98%
4-Fluoro-2-nitrobenzyl bromide
Formula: C₇H₅BrFNO₂
MW: 234.023 g/mol
Density: 1.6894 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07779495
CAS Number: 76437-44-0
UN: 3265
ADR: 8 II