3-Bromo-5-(trifluoromethoxy)benzene-1,2-diamine
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-2,3-diamino-5-(trifluoromethoxy)benzene, 3-Bromo-5-(trifluoromethoxy)phenylene-1,2-diamine
PC48404-1G PC48404-250MG PC48404-5G
APOSPC48404-1GEA 139 EUR
APOSPC48404-1G APOSPC48404-250MG APOSPC48404-5G
3-Bromo-5-(trifluoromethoxy)benzene-1,2-diamine
3-Bromo-5-(trifluoromethoxy)benzene-1,2-diamine
Formula: C₇H₆BrF₃N₂O
MW: 271.035 g/mol
Melting Pt: 39…40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042191
CAS Number: 1191922-50-5