3,3,3-Trifluoropropylene carbonate
Supplier: Apollo Scientific
Warning
Synonyms: 4-(Trifluoromethyl)-1,3-dioxolan-2-one
PC48592-1G PC48592-5G
APOSPC48592-1GEA 21.9 EUR
APOSPC48592-1G APOSPC48592-5G
3,3,3-Trifluoropropylene carbonate
3,3,3-Trifluoropropylene carbonate
Formula: C₄H₃F₃O₃
Boiling Pt: 85…86 °C (12 mm Hg)
Melting Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD02183522
CAS Number: 167951-80-6