3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)benzonitrile
Supplier: Apollo Scientific
PC48630-1G PC48630-250MG PC48630-5G
APOSPC48630-1GEA 244 EUR
APOSPC48630-1G APOSPC48630-250MG APOSPC48630-5G
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)benzonitrile
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)benzonitrile
Formula: C₁₄H₁₅BF₃NO₂
MW: 297.081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12405369
CAS Number: 479411-95-5