3-Bromo-4-(trifluoromethoxy)benzoic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-4-carboxy-α,α,α-trifluoroanisole
PC49212-1G PC49212-5G
APOSPC49212-1GEA 112 EUR
APOSPC49212-1G APOSPC49212-5G
3-Bromo-4-(trifluoromethoxy)benzoic acid 98%
3-Bromo-4-(trifluoromethoxy)benzoic acid
Formula: C₈H₄BrF₃O₃
MW: 285.015 g/mol
Melting Pt: 126.5…126.7 °C
Storage Temperature: Ambient
MDL Number: MFCD11110888
CAS Number: 85373-96-2