4-Bromo-3-(trifluoromethoxy)benzaldehyde 95%
Supplier: Apollo Scientific
Warning
PC49240-1G PC49240-250MG
APOSPC49240-1GEA 174 EUR
APOSPC49240-1G APOSPC49240-250MG
4-Bromo-3-(trifluoromethoxy)benzaldehyde 95%
4-Bromo-3-(trifluoromethoxy)benzaldehyde
Formula: C₈H₄BrF₃O₂
MW: 269.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12924842
CAS Number: 1221716-04-6