Methyl 2-bromo-2-fluoropropanoate 99%
Supplier: Apollo Scientific
|
Warning
|
PC49318-25G
PC49318-5G
APOSPC49318-25GEA
1110
EUR
APOSPC49318-25G
APOSPC49318-5G
Methyl 2-bromo-2-fluoropropanoate 99%
Methyl 2-bromo-2-fluoropropanoate
Formula:
C₄H₆BrFO₂ Boiling Pt: 65…66 °C (75 mm Hg) Density: 1.54 g/cm³ (25 °C) Flash Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD18074353 CAS Number: 157415-07-1 |