6-Bromo-2-hydroxy-3-nitrobenzotrifluoride 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-6-nitro-2-(trifluoromethyl)phenol, 3-Bromo-6-nitro-α,α,α-trifluoro-o-cresol
PC49607-1G PC49607-250MG
APOSPC49607-1GEA 615 EUR
APOSPC49607-1G APOSPC49607-250MG
6-Bromo-2-hydroxy-3-nitrobenzotrifluoride 97%
6-Bromo-2-hydroxy-3-nitrobenzotrifluoride
Formula: C₇H₃BrF₃NO₃
MW: 286.003 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25541885
CAS Number: 1445995-70-9