tert-Butyl 5-(trifluoromethyl)-3,4-dihydro-1,8-naphthyridine-1(2H)-carboxylate 95%
Supplier: Apollo Scientific
Warning
PC49673-1G PC49673-250MG
APOSPC49673-1GEA 111 EUR
APOSPC49673-1G APOSPC49673-250MG
tert-Butyl 5-(trifluoromethyl)-3,4-dihydro-1,8-naphthyridine-1(2H)-carboxylate 95%
tert-Butyl 5-(trifluoromethyl)-3,4-dihydro-1,8-naphthyridine-1(2H)-carboxylate
Formula: C₁₄H₁₇F₃N₂O₂
MW: 302.292 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32066748
CAS Number: 2366994-11-6