2-Fluoro-3-methylphenyl trifluoromethanesulphonate 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Fluoro-3-methylphenyl triflate, 2-Fluoro-3-{[(trifluoromethyl)sulphonyl]oxy}toluene
PC500061-1G PC500061-250MG
APOSPC500061-1GEA 102 EUR
APOSPC500061-1G APOSPC500061-250MG
2-Fluoro-3-methylphenyl trifluoromethanesulphonate 97%
2-Fluoro-3-methylphenyl trifluoromethanesulphonate
Formula: C₈H₆F₄O₃S
MW: 258.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25541823
CAS Number: 1446016-74-5
UN: 3265
ADR: 8 III