3-Methyl-2-nitrophenyl trifluoromethanesulphonate
Supplier: Apollo Scientific
Danger
Synonyms: 3-Methyl-2-nitrophenyl triflate, 2-Nitro-3-{[(trifluoromethyl)sulphonyl]oxy}toluene
PC500110-1G PC500110-250MG
APOSPC500110-1GEA 488 EUR
APOSPC500110-1G APOSPC500110-250MG
3-Methyl-2-nitrophenyl trifluoromethanesulphonate
3-Methyl-2-nitrophenyl trifluoromethanesulphonate
Formula: C₈H₆F₃NO₅S
Melting Pt: 24…26 °C
Storage Temperature: Ambient
MDL Number: MFCD25541870
CAS Number: 145209-21-8
UN: 3261
ADR: 8,III