6-(Bromomethyl)-2,2,3,3-tetrafluoro-1,4-benzodioxane
Supplier: Apollo Scientific
Warning
PC500586-1G PC500586-250MG PC500586-5G
APOSPC500586-1GEA 1350 EUR
APOSPC500586-1G APOSPC500586-250MG APOSPC500586-5G
6-(Bromomethyl)-2,2,3,3-tetrafluoro-1,4-benzodioxane
6-(Bromomethyl)-2,2,3,3-tetrafluoro-1,4-benzodioxane
Formula: C₉H₅BrF₄O₂
MW: 301.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28096597
CAS Number: 1823569-26-1