4-[1,1,2-Trifluoro-2-(trifluoromethoxy)ethoxy]benzoic acid
Supplier: Apollo Scientific
Warning
PC500652-1G PC500652-250MG
APOSPC500652-1GEA 140 EUR
APOSPC500652-1G APOSPC500652-250MG
4-[1,1,2-Trifluoro-2-(trifluoromethoxy)ethoxy]benzoic acid
4-[1,1,2-Trifluoro-2-(trifluoromethoxy)ethoxy]benzoic acid
Formula: C₁₀H₆F₆O₄
MW: 304.143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103420
CAS Number: 1845695-98-8